Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rock2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009072.2 | NP_033098.2 | 9 | 1384 | ||
GenBank mRNA | AK144687.1 | - | 9 | 1238 | ||
AK150181.1 | - | 9 | 1238 | |||
AK163082.1 | - | 9 | 1238 | |||
DQ864977.1 | - | 9 | 1195 | |||
U58513.1 | - | 9 | 1172 |