Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164634.1 | NP_001158106.1 | 10 | 1566 | ||
NM_001164635.1 | NP_001158107.1 | 10 | 1474 | |||
NM_031194.5 | NP_112471.3 | 11 | 1646 | |||
GenBank mRNA | AB079895.1 | - | 9 | 1303 | ||
AF078869.1 | - | 10 | 1421 | |||
AK031962.1 | - | 11 | 1612 | |||
AK044336.1 | - | 10 | 1443 | |||
BC014762.1 | - | 10 | 1452 | 4239544 |