Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rora | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289916.1 | NP_001276845.1 | 4 | 679 | ||
NM_013646.2 | NP_038674.1 | 5 | 688 | |||
GenBank mRNA | AK043990.1 | - | 1 | 48 | ||
AK149294.1 | - | 4 | 569 | |||
AK155827.1 | - | 4 | 630 | |||
BC003757.1 | - | 4 | 628 | 3592667 | ||
S82720.1 | - | 4 | 605 | |||
U53228.1 | - | 5 | 672 | |||
Y08640.1 | - | 4 | 834 | |||
Z82994.1 | - | 4 | 667 |