Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpe65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029987.2 | NP_084263.2 | 5, 6 | 555 | ||
GenBank mRNA | AF410461.1 | - | 4, 5 | 503 | ||
AK020906.1 | - | 5, 6 | 554 | |||
BC090656.1 | - | 5, 6 | 531 | |||
BC130028.1 | - | 4, 5 | 507 | 40140883 | ||
CJ133618.1 | - | 5, 6 | 556 |