Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009077.2 | NP_033103.2 | 2, 3 | 151 | ||
GenBank mRNA | AK007263.1 | - | 2, 3 | 121 | ||
AK009044.1 | - | 2, 3 | 122 | |||
AK010510.1 | - | 2, 3 | 119 | |||
AK012580.1 | - | 2, 3 | 119 | |||
AK145649.1 | - | 2, 3 | 152 | |||
AK151667.1 | - | 2, 3 | 121 | |||
AK153172.1 | - | 2, 3 | 119 | |||
AK166803.1 | - | 2, 3 | 121 | |||
AK168288.1 | - | 2, 3 | 119 | |||
AK169659.1 | - | 2, 3 | 119 | |||
BC072558.1 | - | 2460 | ||||
BC081468.1 | - | 2, 3 | 109 | 6823249 | ||
BC082290.1 | - | 2, 3 | 105 | 6485569 | ||
BC091732.1 | - | 1, 2 | 97 | 2655484 | ||
DQ403025.1 | - | 1, 2 | 16 | |||
L04128.1 | - | 1, 2 | 111 |