Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009091.2 | NP_033117.1 | 2, 3 | 132 | ||
GenBank mRNA | AA027738.1 | - | 1, 2 | 60 | ||
AK135319.1 | - | 2, 3 | 106 | |||
AK146147.1 | - | 2, 3 | 104 | |||
AV569793.1 | - | 2, 3 | 132 | |||
BC010763.1 | - | 1, 2 | 98 | 4234356 | ||
BC094409.1 | - | 1, 2 | 97 | 1179896 | ||
M33330.1 | - | 2, 3 | 112 |