Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013647.2 | NP_038675.2 | 4 | 396 | ||
GenBank mRNA | AK002904.1 | - | 1 | 334 | ||
AK010613.1 | - | 1 | 337 | |||
AK010641.1 | - | 1 | 337 | |||
AK011060.1 | - | 1 | 348 | |||
AK019400.2 | - | 1 | 335 | |||
AK083946.1 | - | 1 | 335 | |||
AK088413.1 | - | 1 | 336 | |||
AK088716.1 | - | 1 | 336 | |||
AK150615.1 | - | 1 | 327 | |||
AK150808.1 | - | 1 | 338 | |||
AK168207.1 | - | 1 | 335 | |||
AK169328.1 | - | 1 | 336 | |||
BC082286.1 | - | 1 | 315 | |||
BC090618.1 | - | 1 | 363 | 5713263 | ||
BC115673.1 | - | 1 | 303 | 40104131 | ||
BC115674.1 | - | 1 | 303 | 40104132 |