Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011297.2 | NP_035427.2 | 6 | 716 | ||
NM_207634.1 | NP_997517.1 | 5 | 696 | |||
NM_207635.1 | NP_997518.1 | 7 | 734 | |||
GenBank mRNA | AK010856.1 | - | 5 | 660 | ||
AK049387.1 | - | 656 |