Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011300.3 | NP_035430.1 | 4 | 285 | ||
GenBank mRNA | AK135520.1 | - | 4 | 214 | ||
AK160632.1 | - | 4 | 214 | |||
AK166096.1 | - | 4 | 214 | |||
AK169636.1 | - | 4 | 214 | |||
AV470458.1 | - | 4 | 285 | |||
BC002014.1 | - | 3 | 204 | 3484169 | ||
BC086785.1 | - | 191 | ||||
BC099605.1 | - | 3 | 181 | 1528652 | ||
BC100397.1 | - | 3 | 200 | 6439354 | ||
BU524389.1 | - | 4 | 276 | |||
EU349428.1 | - | 3 | 178 |
Gene Symbol: Gm6252 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK132285.1 | - | 1 | 713 |