Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011302.3 | NP_035432.3 | 5 | 628 | ||
GenBank mRNA | AF073780.1 | - | 5 | 491 | ||
AF084561.1 | - | 5 | 499 | |||
AJ011381.1 | - | 5 | 479 | |||
BC046422.1 | - | 5 | 628 | 5401012 |