Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003933.2 | NP_001003933.1 | 4 | 2784 | ||
NM_001003934.2 | NP_001003934.1 | 5 | 2841 | |||
NM_001271486.1 | NP_001258415.1 | 5 | 1878 | |||
NM_001271487.1 | NP_001258416.1 | 4 | 717 | |||
NM_053076.3 | NP_444306.1 | 3 | 660 | |||
GenBank mRNA | AB046114.1 | - | 3 | 654 | ||
AF195940.1 | - | 3 | 604 | |||
AK032109.1 | - | 5 | 1855 | |||
AK036892.1 | - | 3 | 626 | |||
AK049845.1 | - | 3 | 592 | |||
AK075883.1 | - | 3 | 589 | |||
AK088670.1 | - | 3 | 592 | |||
AK146505.1 | - | 3 | 468 | |||
AK148792.1 | - | 3 | 619 | |||
AK148947.1 | - | 3 | 621 | |||
AK165683.1 | - | 3 | 591 | |||
AK197640.1 | - | 2 | 310 | |||
AK217412.1 | - | 2 | 217 | |||
AK218971.1 | - | 2 | 226 | |||
AY164700.1 | - | 3 | 654 | |||
AY427822.1 | - | 4 | 2765 | |||
AY507126.1 | - | 3 | 452 | |||
AY507127.1 | - | 3 | 2387 | |||
AY750849.1 | - | 5 | 2822 | |||
BC005478.1 | - | 3 | 563 | |||
BC014697.1 | - | 3 | 571 | |||
BC036717.1 | - | 3 | 569 | |||
BC079882.1 | - | 5 | 2751 | 6839323 |