Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003933.2 | NP_001003933.1 | 6 | 2928 | ||
NM_001003934.2 | NP_001003934.1 | 7 | 2985 | |||
NM_001271486.1 | NP_001258415.1 | 7 | 2022 | |||
NM_001271487.1 | NP_001258416.1 | 6 | 861 | |||
NM_053076.3 | NP_444306.1 | 5 | 804 | |||
GenBank mRNA | AB046114.1 | - | 5 | 798 | ||
AF195940.1 | - | 5 | 748 | |||
AK032109.1 | - | 7 | 1999 | |||
AK036892.1 | - | 5 | 770 | |||
AK049845.1 | - | 5 | 736 | |||
AK075883.1 | - | 5 | 733 | |||
AK088670.1 | - | 5 | 736 | |||
AK146505.1 | - | 5 | 612 | |||
AK148792.1 | - | 5 | 763 | |||
AK148947.1 | - | 5 | 765 | |||
AK165683.1 | - | 5 | 735 | |||
AK194472.1 | - | 108 | ||||
AK218971.1 | - | 4 | 370 | |||
AY164700.1 | - | 5 | 798 | |||
AY427822.1 | - | 6 | 2909 | |||
AY507126.1 | - | 5 | 596 | |||
AY507127.1 | - | 5 | 2531 | |||
AY750849.1 | - | 7 | 2966 | |||
BC005478.1 | - | 5 | 707 | |||
BC014697.1 | - | 5 | 715 | |||
BC036717.1 | - | 5 | 713 | |||
BC079882.1 | - | 7 | 2895 | 6839323 |