Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uimc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293660.1 | NP_001280589.1 | 8 | 872 | ||
NM_011307.2 | NP_035437.1 | 10 | 1710 | |||
GenBank mRNA | AK052704.1 | - | 10 | 1711 | ||
AK083342.1 | - | 10 | 1651 | |||
AK160926.1 | - | 8 | 873 | |||
BC037092.1 | - | 9 | 1585 | |||
BC040808.1 | - | 10 | 1666 | 4038778 | ||
BC051659.1 | - | 10 | 1633 | |||
BC066220.1 | - | 10 | 1633 | |||
U22015.1 | - | 6 | 1162 |