Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ryk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042607.1 | NP_001036072.1 | 14 | 1761 | ||
NM_001284258.1 | NP_001271187.1 | 13 | 1639 | |||
NM_013649.3 | NP_038677.3 | 14 | 1770 | |||
GenBank mRNA | AK076449.1 | - | 8 | 860 | ||
AK144355.1 | - | 13 | 1579 | |||
BC006963.1 | - | 10 | 1069 | |||
BC032275.1 | - | 14 | 1495 | 5370100 | ||
L02210.1 | - | 14 | 1745 | |||
L21707.1 | - | 14 | 1779 | |||
L38394.1 | - | 14 | 1693 | |||
M98547.1 | - | 14 | 1734 |