Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ryr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023868.2 | NP_076357.2 | 37 | 5663 | ||
GenBank mRNA | AF295105.1 | - | 37 | 5164 | ||
BU921357.1 | - | 2 | 441 |