Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S100a13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009113.4 | NP_033139.2 | 3, 4 | 497 | ||
GenBank mRNA | AK002755.1 | - | 2, 3 | 296 | ||
AK008895.1 | - | 2, 3 | 296 | |||
BC005687.1 | - | 3, 4 | 497 | 3155915 | ||
CK334302.1 | - | 2, 3 | 464 | |||
CX219167.1 | - | 2, 3 | 220 | |||
X99921.1 | - | 2, 3 | 224 |