Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S100a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011311.2 | NP_035441.1 | 3 | 227 | ||
GenBank mRNA | AK003065.1 | - | 3 | 233 | ||
AK160522.1 | - | 3 | 228 | |||
BC051214.1 | - | 2 | 186 | 1383390 | ||
BC094635.1 | - | 2 | 186 | 1248264 | ||
BX526220.1 | - | 2 | 178 | |||
D00208.1 | - | 3 | 227 | |||
X05835.1 | - | 2 | 184 | |||
X16190.1 | - | 2 | 186 | |||
Z36947.1 | - | 481 |