Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Saa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009117.3 | NP_033143.1 | 4 | 308 | ||
GenBank mRNA | AK136475.1 | - | 4 | 314 | ||
BC052495.1 | - | 1 | 2150 | |||
BC087933.1 | - | 4 | 308 | 30310171 |