Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Saa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011316.3 | NP_035446.1 | 3 | 244 | ||
GenBank mRNA | BC019212.1 | - | 3 | 280 | 5053373 | |
U02554.1 | - | 3 | 275 |