Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004521.2 | NP_004512.1 | 20, 21 | 2771 | ||
XM_017016224.1 | XP_016871713.1 | 20, 21 | 2377 | |||
GenBank mRNA | AK289413.1 | - | 20, 21 | 2717 | ||
AK299542.1 | - | 1142 | ||||
BC126279.1 | - | 20, 21 | 2524 | 8991995 | ||
BC126281.1 | - | 20, 21 | 2524 | 8991997 | ||
FJ224324.1 | - | 20, 21 | 2614 | |||
X65873.1 | - | 20, 21 | 2614 |