Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sart1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016882.3 | NP_058578.3 | 6 | 785 | ||
GenBank mRNA | AB014721.1 | - | 6 | 743 | ||
AF129931.1 | - | 6 | 785 | |||
AK088341.1 | - | 6 | 745 | |||
BC028823.1 | - | 6 | 747 | 5097832 | ||
BC051394.1 | - | 6 | 747 | 5097832 |