Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nkx1-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009123.2 | NP_033149.1 | 1 | 188 | ||
GenBank mRNA | AF222444.1 | - | 1 | 188 | ||
AF222445.1 | - | 1 | 189 | |||
AF223361.1 | - | 1 | 190 | |||
AF223362.1 | - | 1 | 189 | |||
AF223363.1 | - | 1 | 188 | |||
AF223674.1 | - | 1 | 190 | |||
AF223675.1 | - | 1 | 190 | |||
BC139757.1 | - | 1 | 117 | 40047346 | ||
X75384.1 | - | 1 | 293 |