Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009127.4 | NP_033153.2 | 2, 3 | 606 | ||
GenBank mRNA | AF509567.1 | - | 2, 3 | 441 | ||
AF509570.1 | - | 2, 3 | 441 | |||
AK142630.1 | - | 2, 3 | 459 | |||
AK148103.1 | - | 2, 3 | 569 | |||
BC007474.1 | - | 2, 3 | 450 | 3599932 | ||
BC055453.1 | - | 2, 3 | 564 | 4458907 | ||
CT010171.1 | - | 2, 3 | 306 | |||
CT010332.1 | - | 2, 3 | 306 |