Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146196.1 | NP_001139668.1 | 3 | 520 | ||
NM_009132.2 | NP_033158.2 | 3 | 520 | |||
GenBank mRNA | AK133484.1 | - | 3 | 476 | ||
AK136080.1 | - | 1 | 62 | |||
AK167739.1 | - | 3 | 475 | |||
AK171530.1 | - | 3 | 507 | |||
AK173295.1 | - | 3 | 482 | |||
BC063328.1 | - | 3 | 475 | 30314464 | ||
U04354.1 | - | 3 | 475 | |||
Y13971.1 | - | 3 | 475 |