Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXCR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168298.1 | NP_001161770.1 | 4 | 838 | ||
NM_001557.3 | NP_001548.1 | 3 | 1046 | |||
XM_005246530.3 | XP_005246587.1 | 4 | 818 | |||
XM_017003990.1 | XP_016859479.1 | 4 | 798 | |||
XM_017003991.1 | XP_016859480.1 | 2 | 709 | |||
XM_017003992.1 | XP_016859481.1 | 2 | 979 | |||
GenBank mRNA | AK290906.1 | - | 3 | 944 | ||
AK312664.1 | - | 3 | 937 | |||
BC037961.1 | - | 3 | 1000 | 5752441 | ||
L19593.1 | - | 3 | 1036 | |||
M73969.1 | - | 1 | 653 | |||
M94582.1 | - | 2 | 680 |