Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL9R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002186.2 | NP_002177.2 | 5 | 711 | ||
NM_176786.1 | NP_789743.2 | 6 | 800 | |||
XM_011531151.2 | XP_011529453.1 | 6 | 761 | |||
XM_011531152.2 | XP_011529454.1 | 6 | 737 | |||
XM_011531154.2 | XP_011529456.1 | 6 | 783 | |||
XM_011531155.2 | XP_011529457.1 | 6 | 770 | |||
XM_011531156.2 | XP_011529458.1 | 6 | 768 | |||
XM_011531157.2 | XP_011529459.1 | 6 | 765 | |||
XM_017029495.1 | XP_016884984.1 | 6 | 763 | |||
XM_017029496.1 | XP_016884985.1 | 6 | 771 | |||
XM_017029497.1 | XP_016884986.1 | 6 | 807 | |||
XM_017029498.1 | XP_016884987.1 | 7 | 912 | |||
XM_017029499.1 | XP_016884988.1 | 7 | 834 | |||
XM_017029500.1 | XP_016884989.1 | 6 | 774 | |||
XM_017029501.1 | XP_016884990.1 | 6 | 764 | |||
XM_017029502.1 | XP_016884991.1 | 2 | 214 | |||
XM_017029503.1 | XP_016884992.1 | 7 | 813 | |||
XM_017029504.1 | XP_016884993.1 | 6 | 786 | |||
XM_017029505.1 | XP_016884994.1 | 6 | 768 | |||
GenBank mRNA | AK303178.1 | - | 6 | 775 | ||
BC051337.1 | - | 6 | 675 | 6304829 | ||
M84747.1 | - | 5 | 711 |