Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL9R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002186.2 | NP_002177.2 | 9 | 1976 | ||
NM_176786.1 | NP_789743.2 | 9 | 1980 | |||
XM_011531151.2 | XP_011529453.1 | 10 | 2026 | |||
XM_011531152.2 | XP_011529454.1 | 10 | 2002 | |||
XM_011531154.2 | XP_011529456.1 | 10 | 2048 | |||
XM_017029495.1 | XP_016884984.1 | 10 | 2025 | |||
XM_017029496.1 | XP_016884985.1 | 9 | 1961 | |||
XM_017029497.1 | XP_016884986.1 | 10 | 2072 | |||
XM_017029498.1 | XP_016884987.1 | 11 | 2177 | |||
XM_017029499.1 | XP_016884988.1 | 11 | 2099 | |||
XM_017029501.1 | XP_016884990.1 | 7 | 1636 | |||
XM_017029502.1 | XP_016884991.1 | 6 | 1479 | |||
XM_017029506.1 | XP_016884995.1 | 3 | 1394 | |||
GenBank mRNA | BC051337.1 | - | 9 | 1855 | 6304829 |