Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3h7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081016.1 | NP_001074485.1 | 13, 14 | 1973 | ||
GenBank mRNA | AK129268.1 | - | 13, 14 | 1650 | ||
AK147484.1 | - | 13, 14 | 1737 | |||
AK147978.1 | - | 13, 14 | 1746 | |||
AK157711.1 | - | 5, 6 | 1343 | |||
AK171949.1 | - | 13, 14 | 1688 | |||
BC059923.1 | - | 13, 14 | 1728 |