Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sct | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287171.1 | NP_001274100.1 | 3 | 296 | ||
NM_011328.3 | NP_035458.1 | 3 | 296 | |||
GenBank mRNA | AK145859.1 | - | 3 | 295 | ||
BC048484.1 | - | 3 | 296 | 6707701 | ||
CF616255.1 | - | 267 | ||||
X73580.1 | - | 3 | 269 |