Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL10RA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001558.3 | NP_001549.2 | 7 | 1518 | ||
NR_026691.1 | 8 | 1648 | ||||
GenBank mRNA | AB209626.1 | - | 6 | 3019 | ||
AK291645.1 | - | 7 | 1518 | |||
AK301260.1 | - | 6 | 1336 | |||
AK301484.1 | - | 8 | 1648 | |||
AK301492.1 | - | 7 | 1788 | |||
AK313049.1 | - | 7 | 1512 | |||
AK316146.1 | - | 7 | 1570 | |||
AK316342.1 | - | 8 | 1722 | |||
BC028082.1 | - | 7 | 1495 | 5216986 | ||
JN591377.1 | - | 7 | 1441 | |||
U00672.1 | - | 7 | 1502 |