Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccl7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013654.3 | NP_038682.1 | 2 | 212 | ||
GenBank mRNA | AF128193.1 | - | 2 | 177 | ||
AF128194.1 | - | 2 | 177 | |||
AK078827.1 | - | 2 | 213 | |||
BC061126.1 | - | 2 | 210 | 30276350 | ||
BY163435.1 | - | 2 | 214 | |||
L04694.1 | - | 2 | 174 | |||
S50588.1 | - | 2 | 160 | |||
S71251.1 | - | 2 | 192 | |||
Z12297.1 | - | 2 | 160 |