Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014418.1 | - | 4 | 674 | ||
AK157860.1 | - | 4 | 539 | |||
AK163400.1 | - | 4 | 659 | |||
AK167113.1 | - | 4 | 588 | |||
BC005715.1 | - | 5 | 808 | |||
BC068152.1 | - | 4 | 914 | 30534982 | ||
D50461.1 | - | 4 | 917 | |||
U45977.1 | - | 4 | 822 | |||
U45978.1 | - | 4 | 690 |