Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nptn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293673.1 | NP_001280602.1 | 5 | 738 | ||
NM_009145.2 | NP_033171.2 | 5 | 738 | |||
GenBank mRNA | AK075610.1 | - | 5 | 573 | ||
AK076624.1 | - | 338 | ||||
AK153930.1 | - | 5 | 571 | |||
BC039191.1 | - | 5 | 588 | |||
D50463.1 | - | 5 | 569 |