Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exoc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009148.3 | NP_033174.2 | 10 | 1505 | ||
GenBank mRNA | AF022962.1 | - | 10 | 1531 | ||
AK028250.1 | - | 8 | 1237 | |||
AK039983.1 | - | 10 | 1528 | |||
AK086603.1 | - | 10 | 1524 | |||
AK173235.1 | - | 10 | 1508 | |||
BC034644.1 | - | 10 | 1505 | 5355276 |