Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Selenbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009150.3 | NP_033176.2 | 3 | 221 | ||
GenBank mRNA | AK002447.1 | - | 3 | 239 | ||
BC011202.1 | - | 3 | 221 | 4209393 | ||
M32032.1 | - | 204 |
Gene Symbol: Selenbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019414.2 | NP_062287.1 | 3 | 231 | ||
GenBank mRNA | BC024106.1 | - | 3 | 229 | 5096831 | |
S56599.1 | - | 3 | 231 |