Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Selenbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019414.2 | NP_062287.1 | 8, 9 | 1011 | ||
GenBank mRNA | BC024106.1 | - | 8, 9 | 1009 | 5096831 | |
S56599.1 | - | 8, 9 | 1011 |
Gene Symbol: Selenbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009150.3 | NP_033176.2 | 8, 9 | 1001 | ||
GenBank mRNA | AK002447.1 | - | 8, 9 | 1017 | ||
BC011202.1 | - | 8, 9 | 1001 | 4209393 |