Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema3f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011349.3 | NP_035479.2 | 7 | 778 | ||
GenBank mRNA | AF036162.1 | - | 5 | 436 | ||
AF080090.1 | - | 7 | 684 | |||
AF080091.1 | - | 6 | 577 | |||
AK053632.1 | - | 7 | 779 | |||
AK140340.1 | - | 7 | 1266 | |||
BC010976.1 | - | 7 | 699 | 4219060 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK172610.1 | - | 1171 | Chr. - - [-] on Build |