Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK090073.1 | - | 15 | 2679 | ||
AK148298.1 | - | 15 | 2676 | |||
AK173247.1 | - | 16 | 2917 | |||
CB247850.1 | - | 1 | 380 | |||
S79463.1 | - | 15 | 2531 |