Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema4d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281880.1 | NP_001268809.1 | 17 | 2715 | ||
NM_013660.4 | NP_038688.2 | 16 | 2651 | |||
GenBank mRNA | AK088653.1 | - | 13 | 1933 | ||
AK132688.1 | - | 17 | 2641 | |||
AK150911.1 | - | 15 | 2544 | |||
BC049780.1 | - | 16 | 2548 | 6509473 | ||
U69535.1 | - | 14 | 2178 |