Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009154.2 | NP_033180.2 | 7, 8 | 1088 | ||
GenBank mRNA | AK031231.1 | - | 7, 8 | 1081 | ||
AK043386.1 | - | 7, 8 | 800 | |||
AK045236.1 | - | 7, 8 | 1092 | |||
AK046502.1 | - | 7, 8 | 864 | |||
AK141612.1 | - | 7, 8 | 1032 | |||
AK143026.1 | - | 7, 8 | 1099 | |||
AK157706.1 | - | 7, 8 | 1071 | |||
BC065137.1 | - | 6, 7 | 557 | 30548165 | ||
X97817.1 | - | 5, 6 | 457 |