Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sept8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252332.1 | NP_001239261.1 | 3 | 449 | ||
NM_001252333.1 | NP_001239262.1 | 3 | 449 | |||
NM_033144.2 | NP_149156.1 | 3 | 449 | |||
GenBank mRNA | AF179996.1 | - | 2 | 145 | ||
AK144178.1 | - | 3 | 291 | |||
BC037707.1 | - | 3 | 237 | |||
BC049819.1 | - | 3 | 264 | |||
BC059248.1 | - | 3 | 385 | 6400429 | ||
CJ130914.1 | - | 3 | 449 |