Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sepp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042613.1 | NP_001036078.1 | 5 | 698 | ||
NM_001042614.1 | NP_001036079.1 | 5 | 781 | |||
NM_009155.3 | NP_033181.3 | 5 | 715 | |||
GenBank mRNA | AK002450.1 | - | 5 | 697 | ||
AK077265.1 | - | 5 | 697 | |||
AK146774.1 | - | 5 | 697 | |||
AK148623.1 | - | 5 | 697 | |||
AK159269.1 | - | 5 | 697 | |||
AK159524.1 | - | 5 | 698 | |||
AK159894.1 | - | 5 | 694 | |||
AK159911.1 | - | 5 | 698 | |||
AK159935.1 | - | 5 | 697 | |||
AK167611.1 | - | 5 | 697 | |||
AK168463.1 | - | 5 | 694 | |||
AK168583.1 | - | 5 | 694 | |||
AK168631.1 | - | 5 | 697 | |||
AK168747.1 | - | 5 | 697 | |||
AK168926.1 | - | 5 | 697 | |||
AK169011.1 | - | 5 | 696 | |||
AK169158.1 | - | 5 | 696 | |||
AK169362.1 | - | 5 | 697 | |||
AK183252.1 | - | 1 | 34 | |||
AK190256.1 | - | 1 | 34 | |||
AK199458.1 | - | 1 | 34 | |||
BC001991.1 | - | 5 | 698 | 3486761 | ||
X99807.1 | - | 4 | 758 |