Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL12B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002187.2 | NP_002178.2 | 3 | 394 | ||
GenBank mRNA | AF180563.1 | - | 2 | 358 | ||
BC067498.1 | - | 2 | 356 | 6971868 | ||
BC067499.1 | - | 2 | 356 | 6971869 | ||
BC067500.1 | - | 2 | 356 | 6971872 | ||
BC067501.1 | - | 2 | 356 | 6971873 | ||
BC067502.1 | - | 2 | 356 | 6971874 | ||
BC074723.2 | - | 3 | 383 | 30915173 | ||
M65272.1 | - | 3 | 394 | |||
M65290.1 | - | 2 | 365 |