Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sftpb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282071.2 | NP_001269000.1 | 4 | 311 | ||
NM_147779.2 | NP_680088.1 | 4 | 311 | |||
GenBank mRNA | AI894333.1 | - | 1 | 40 |