Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sftpd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009160.2 | NP_033186.1 | 4 | 478 | ||
GenBank mRNA | BC003705.1 | - | 4 | 429 | 3601620 | |
BC012896.1 | - | 3 | 666 | |||
L40156.1 | - | 3 | 406 |