Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081459.2 | NP_001074928.1 | 3 | 1568 | ||
NM_001289538.1 | NP_001276467.1 | 3 | 1568 | |||
NM_001289539.1 | NP_001276468.1 | 3 | 1191 | |||
NM_001289540.1 | NP_001276469.1 | 4 | 1328 | |||
NM_001289541.1 | NP_001276470.1 | 3 | 1191 | |||
NM_001289542.1 | NP_001276471.1 | 3 | 1568 | |||
NM_011363.3 | NP_035493.2 | 3 | 1568 | |||
GenBank mRNA | AF020526.1 | - | 3 | 1262 | ||
AF074329.1 | - | 2 | 973 | |||
AF380422.1 | - | 3 | 1262 | |||
AF421138.1 | - | 3 | 1262 | |||
AF421139.1 | - | 3 | 1262 | |||
AK148085.1 | - | 4 | 1295 | |||
AK154655.1 | - | 3 | 1502 | |||
AK168439.1 | - | 3 | 1163 | |||
AK170444.1 | - | 3 | 1542 | |||
AK171328.1 | - | 3 | 1576 | |||
AK173145.1 | - | 2 | 715 | |||
BC011422.1 | - | 2 | 1030 | 3156603 | ||
BC051978.1 | - | 3 | 1111 | 6401205 |