Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3gl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277954.1 | NP_001264883.1 | 3 | 351 | ||
NM_001277955.1 | NP_001264884.1 | 3 | 351 | |||
NM_017400.6 | NP_059096.3 | 3 | 351 | |||
GenBank mRNA | AK012114.1 | - | 293 | |||
AK157464.1 | - | 305 | ||||
BC027096.1 | - | 351 | 5254139 | |||
BC100453.1 | - | 196 | 6770826 | |||
BE653420.1 | - | 203 | ||||
U58887.1 | - | 331 |