Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sorbs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034962.1 | NP_001030134.1 | 7 | 552 | ||
NM_001034963.1 | NP_001030135.1 | 6 | 462 | |||
NM_001034964.1 | NP_001030136.1 | 5 | 377 | |||
NM_009166.3 | NP_033192.2 | 8 | 741 | |||
NM_178362.1 | NP_848139.1 | 8 | 1200 | |||
GenBank mRNA | AF078666.1 | - | 6 | 451 | ||
AF078667.1 | - | 7 | 541 | |||
AF521593.1 | - | 4 | 334 | |||
AK035212.1 | - | 8 | 916 | |||
AK087553.1 | - | 8 | 1202 | |||
AK122487.2 | - | 8 | 523 | |||
AK146712.1 | - | 5 | 380 | |||
AK177559.1 | - | 2 | 69 | |||
AK208107.1 | - | 3 | 206 | |||
AK216997.1 | - | 3 | 232 | |||
BC012703.1 | - | 8 | 574 | 3602997 | ||
U58883.1 | - | 8 | 679 |