Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009171.2 | NP_033197.2 | 7 | 1159 | ||
GenBank mRNA | AF237702.1 | - | 7 | 985 | ||
AK010439.1 | - | 7 | 1055 | |||
AK132342.1 | - | 1030 | ||||
BC026055.1 | - | 1024 | 4160579 | |||
BQ832130.1 | - | 2 | 271 | |||
X94478.1 | - | 7 | 985 | |||
X94479.1 | - | 7 | 985 |