Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyfip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164661.1 | NP_001158133.1 | 28 | 3384 | ||
NM_001164662.1 | NP_001158134.1 | 27 | 3325 | |||
NM_011370.3 | NP_035500.2 | 27 | 3331 | |||
GenBank mRNA | AF072697.1 | - | 27 | 3126 | ||
AJ567911.1 | - | 26 | 3093 | |||
AK028811.1 | - | 18 | 2123 | |||
AK044175.1 | - | 27 | 3153 | |||
AK128952.1 | - | 27 | 3118 | |||
AK146613.1 | - | 27 | 3134 | |||
AK152558.1 | - | 28 | 3200 | |||
AK157773.1 | - | 27 | 3119 | |||
AK167068.1 | - | 27 | 3155 | |||
AK178624.1 | - | 2 | 104 | |||
AK196585.1 | - | 2 | 209 | |||
AK198587.1 | - | 2 | 102 | |||
AK220320.1 | - | 27 | 3172 | |||
BC047135.1 | - | 27 | 3120 | 6308554 | ||
BC052713.1 | - | 27 | 3190 | 6835403 | ||
BC054429.1 | - | 26 | 3104 | 5345535 | ||
BC086625.1 | - | 11 | 4106 |